Product Details

SNP ID
rs60535349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99758213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGTTGAAGAAGTCCTCCTAAGC[A/T]TAATTTCAGGGGGATCTGCAACAGT
Phenotype
MIM: 124010
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CYP3A4 PubMed Links

Gene Details

Gene
CYP3A4
Gene Name
cytochrome P450 family 3 subfamily A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202855.2 1536 Missense Mutation AGC,TGC S477C NP_001189784.1
NM_017460.5 1536 Missense Mutation AGC,TGC S478C NP_059488.2
XM_017011780.1 1536 Intron XP_016867269.1

View Full Product Details