Product Details

SNP ID
rs56321035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114789209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAGGTTTCTTCAGGGAACTAAT[C/T]AGCTGGCTATCTTCCAGTCAGCTGG
Phenotype
MIM: 604052
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNFSF15 PubMed Links

Gene Details

Gene
TNFSF15
Gene Name
tumor necrosis factor superfamily member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204344.1 2115 UTR 3 NP_001191273.1
NM_005118.3 2115 UTR 3 NP_005109.2

View Full Product Details