Product Details

SNP ID
rs58280857
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127506076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGAGGGAGGCAGGCTGGGGCC[A/G]GGAGAAGGGGATACACAGGCCCATA
Phenotype
MIM: 614045 MIM: 610933
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
FAM129B PubMed Links
Additional Information
For this assay, SNP(s) [rs11545940] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
FAM129B
Gene Name
family with sequence similarity 129 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035534.2 3041 UTR 3 NP_001030611.1
NM_022833.3 3041 UTR 3 NP_073744.2
XM_005252135.2 3041 UTR 3 XP_005252192.2
XM_011518925.1 3041 UTR 3 XP_011517227.1
Gene
LRSAM1
Gene Name
leucine rich repeat and sterile alpha motif containing 1
There are no transcripts associated with this gene.

View Full Product Details