Product Details

SNP ID
rs61863383
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:62376867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTAAACAGACAGGTGGACGTGG[C/T]CGTGGAAATGATAGCTGTACTGAGG
Phenotype
MIM: 607818
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC283045 PubMed Links

Gene Details

Gene
LOC283045
Gene Name
uncharacterized LOC283045
There are no transcripts associated with this gene.

Gene
ZNF365
Gene Name
zinc finger protein 365
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014951.2 1537 Missense Mutation GCC,GTC A225V NP_055766.2
NM_199450.2 1537 Missense Mutation GCC,GTC A225V NP_955522.1
NM_199451.2 1537 Missense Mutation GCC,GTC A225V NP_955523.1
NM_199452.3 1537 Intron NP_955524.3
XM_017015937.1 1537 Missense Mutation GCC,GTC A225V XP_016871426.1
XM_017015938.1 1537 Intron XP_016871427.1
XM_017015939.1 1537 Intron XP_016871428.1
XM_017015940.1 1537 Intron XP_016871429.1
XM_017015941.1 1537 Intron XP_016871430.1

View Full Product Details