Product Details

SNP ID
rs57734913
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:71749122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTTGTCTCTGCCTCAAGGTCAG[A/C]AGCTGAGGGCTGGGGCCAGGCCAGC
Phenotype
MIM: 615608 MIM: 605516
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
C10orf54 PubMed Links

Gene Details

Gene
C10orf54
Gene Name
chromosome 10 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022153.1 3208 UTR 3 NP_071436.1
Gene
CDH23
Gene Name
cadherin-related 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171930.1 3208 Intron NP_001165401.1
NM_001171931.1 3208 Intron NP_001165402.1
NM_001171932.1 3208 Intron NP_001165403.1
NM_001171933.1 3208 Intron NP_001165404.1
NM_001171934.1 3208 Intron NP_001165405.1
NM_001171935.1 3208 Intron NP_001165406.1
NM_001171936.1 3208 Intron NP_001165407.1
NM_022124.5 3208 Intron NP_071407.4
NM_052836.3 3208 Intron NP_443068.1
XM_006717940.3 3208 Intron XP_006718003.1
XM_006717942.3 3208 Intron XP_006718005.1
XM_011540039.2 3208 Intron XP_011538341.1
XM_011540042.2 3208 Intron XP_011538344.1
XM_011540043.2 3208 Intron XP_011538345.1
XM_011540044.2 3208 Intron XP_011538346.1
XM_011540045.2 3208 Intron XP_011538347.1
XM_011540046.2 3208 Intron XP_011538348.1
XM_011540047.2 3208 Intron XP_011538349.1
XM_011540048.2 3208 Intron XP_011538350.1
XM_011540049.2 3208 Intron XP_011538351.1
XM_011540051.2 3208 Intron XP_011538353.1
XM_011540052.2 3208 Intron XP_011538354.1
XM_017016499.1 3208 Intron XP_016871988.1
XM_017016500.1 3208 Intron XP_016871989.1
XM_017016501.1 3208 Intron XP_016871990.1
XM_017016502.1 3208 Intron XP_016871991.1
XM_017016503.1 3208 Intron XP_016871992.1
XM_017016504.1 3208 Intron XP_016871993.1
XM_017016505.1 3208 Intron XP_016871994.1
XM_017016506.1 3208 Intron XP_016871995.1
XM_017016507.1 3208 Intron XP_016871996.1
XM_017016508.1 3208 Intron XP_016871997.1

View Full Product Details