Product Details

SNP ID
rs61051048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49134153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGCGGATAAATTTAAAGCTGGC[A/G]AATCCTCGGCAAATTTGACTTTTCC
Phenotype
MIM: 111730
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B4GALNT2 PubMed Links

Gene Details

Gene
B4GALNT2
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159387.1 Intron NP_001152859.1
NM_001159388.1 Intron NP_001152860.1
NM_153446.2 Intron NP_703147.2
XM_011524314.2 Intron XP_011522616.1
XM_017024173.1 Intron XP_016879662.1

View Full Product Details