Product Details

SNP ID
rs57775424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:89238440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAGGCAGAAAGGCTGGAGCTATA[C/T]AGGGTACACCTGATGGCTCTGGGTA
Phenotype
MIM: 611241
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPRIN3 PubMed Links

Gene Details

Gene
GPRIN3
Gene Name
GPRIN family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198281.2 11988 Intron NP_938022.2
XM_005262936.3 11988 UTR 3 XP_005262993.1
XM_005262937.4 11988 UTR 3 XP_005262994.1
XM_017008043.1 11988 UTR 3 XP_016863532.1
XM_017008044.1 11988 UTR 3 XP_016863533.1

View Full Product Details