Product Details
- SNP ID
-
rs56094122
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:134909943 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTGAAGCATGACAGTCGGCGTAC[C/T]ACCAGGCTCCTTGGAACTTCTCAGC
- Phenotype
-
MIM: 601252
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FCN1
PubMed Links
Gene Details
- Gene
- FCN1
- Gene Name
- ficolin 1
There are no transcripts associated with this gene.
View Full Product Details