Product Details

SNP ID
rs62244398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:31982529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGCCCAACCCCTTTCTAGCTTCA[C/T]ATCCTTCCAGGAAGATTTCATGGGT
Phenotype
MIM: 606738
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OSBPL10 PubMed Links

Gene Details

Gene
OSBPL10
Gene Name
oxysterol binding protein like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174060.1 Intron NP_001167531.1
NM_017784.4 Intron NP_060254.2
XM_005264843.3 Intron XP_005264900.1
XM_005264844.4 Intron XP_005264901.1
XM_005264845.1 Intron XP_005264902.1
XM_011533326.2 Intron XP_011531628.1
XM_017005667.1 Intron XP_016861156.1
XM_017005668.1 Intron XP_016861157.1
XM_017005669.1 Intron XP_016861158.1
XM_017005670.1 Intron XP_016861159.1
Gene
ZNF860
Gene Name
zinc finger protein 860
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137674.2 Intron NP_001131146.2
XM_017006299.1 Intron XP_016861788.1

View Full Product Details