Product Details
- SNP ID
-
rs62433593
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:154405505 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTAGCAAACTGTGTACAATTTTGA[A/C]GAAGTGTACACACCGTGCTGGAAGA
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CNKSR3
PubMed Links
Gene Details
- Gene
- CNKSR3
- Gene Name
- CNKSR family member 3
View Full Product Details