Product Details

SNP ID
rs56317356
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:51992157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTTGTGCCTTGAATCATTTTC[C/T]CATTGATTTAGTTTTAGTAACACTT
Phenotype
MIM: 610858
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C14orf166 PubMed Links

Gene Details

Gene
C14orf166
Gene Name
chromosome 14 open reading frame 166
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016039.2 Intron NP_057123.1

View Full Product Details