Product Details

SNP ID
rs62004509
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:57564070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATACTGTCGTTTGAGTGTACAG[G/A]TAGTGAGAAAATTTTGTTATATTCA
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
SLC35F4 PubMed Links

Gene Details

Gene
SLC35F4
Gene Name
solute carrier family 35 member F4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206920.1 1837 UTR 3 NP_001193849.1
NM_001306087.1 1837 UTR 3 NP_001293016.1
XM_011536720.2 1837 UTR 3 XP_011535022.1
XM_011536721.2 1837 UTR 3 XP_011535023.1
XM_011536723.2 1837 UTR 3 XP_011535025.1
XM_011536724.2 1837 UTR 3 XP_011535026.1
XM_011536725.1 1837 UTR 3 XP_011535027.1
XM_017021258.1 1837 UTR 3 XP_016876747.1
XM_017021259.1 1837 Intron XP_016876748.1
XM_017021260.1 1837 UTR 3 XP_016876749.1
XM_017021261.1 1837 Intron XP_016876750.1

View Full Product Details