Product Details

SNP ID
rs55784242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:148151591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCAGTTTTTTAATGCCATGTACT[A/C]ATCTAGCTGGCTTAATCTAAAAATA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SPINK14 PubMed Links

Gene Details

Gene
SPINK14
Gene Name
serine peptidase inhibitor, Kazal type 14 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001325.2 Intron NP_001001325.1
XM_017009466.1 Intron XP_016864955.1
XM_017009467.1 Intron XP_016864956.1
XM_017009468.1 Intron XP_016864957.1
XM_017009469.1 Intron XP_016864958.1

View Full Product Details