Product Details

SNP ID
rs60411920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89416858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTCCTAGTCAGGATCAGTACTC[A/G]TAATACAAAAATGGAATTTTCATAT
Phenotype
MIM: 616135
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFIT5 PubMed Links

Gene Details

Gene
IFIT5
Gene Name
interferon induced protein with tetratricopeptide repeats 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012420.2 Intron NP_036552.1

View Full Product Details