Product Details

SNP ID
rs61292440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74262188 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAAAAACACTGTCACCACGCAGG[C/T]ATCTAGTGCCTAGAATCCACTCCTA
Phenotype
MIM: 608987 MIM: 611117
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
P4HA3 PubMed Links

Gene Details

Gene
P4HA3
Gene Name
prolyl 4-hydroxylase subunit alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288748.1 Intron NP_001275677.1
NM_182904.4 Intron NP_878907.1
Gene
PPME1
Gene Name
protein phosphatase methylesterase 1
There are no transcripts associated with this gene.

View Full Product Details