Product Details

SNP ID
rs72634803
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:1334752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGAGGCCCGGAGGGCTCCCAGG[G/A]TACCCGCAACCCACACCGTGAGCTC
Phenotype
MIM: 615467 MIM: 601365 MIM: 605865
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CPTP PubMed Links

Gene Details

Gene
CPTP
Gene Name
ceramide-1-phosphate transfer protein
There are no transcripts associated with this gene.

Gene
DVL1
Gene Name
dishevelled segment polarity protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004421.2 2847 Intron NP_004412.2
XM_005244731.3 2847 Intron XP_005244788.1
XM_005244732.3 2847 Intron XP_005244789.1
XM_005244733.3 2847 Intron XP_005244790.1
Gene
MIR6808
Gene Name
microRNA 6808
There are no transcripts associated with this gene.

Gene
TAS1R3
Gene Name
taste 1 receptor member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152228.2 2847 UTR 3 NP_689414.1
XM_017002435.1 2847 UTR 3 XP_016857924.1
XM_017002436.1 2847 UTR 3 XP_016857925.1

View Full Product Details