Product Details

SNP ID
rs113027410
Assay Type
Functionally tested
NCBI dbSNP Submissions
27
Location
Chr.1:247123549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGGGATCACACACGTATCGAACA[C/T]TGGTTTCCTAAATTAGCAAATCGAC
Phenotype
MIM: 194631
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF124 PubMed Links

Gene Details

Gene
ZNF124
Gene Name
zinc finger protein 124
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243740.2 681 UTR 3 NP_001230669.1
NM_001297567.1 681 Intron NP_001284496.1
NM_001297568.1 681 Intron NP_001284497.1
NM_001297569.1 681 Intron NP_001284498.1
NM_003431.3 681 Intron NP_003422.2
XM_005273257.4 681 Intron XP_005273314.1
XM_006711813.3 681 Intron XP_006711876.1
XM_011544272.2 681 Intron XP_011542574.1

View Full Product Details