Product Details

SNP ID
rs73883902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:180614596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAGAAATTTCTTCTTGAAAGATT[G/A]TTACATTAGAAGTGAGTGTAGTTTC
Phenotype
MIM: 613798
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CCDC39 PubMed Links

Gene Details

Gene
CCDC39
Gene Name
coiled-coil domain containing 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181426.1 3266 UTR 3 NP_852091.1
Gene
TTC14
Gene Name
tetratricopeptide repeat domain 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042601.2 3266 Intron NP_001036066.1
NM_001288582.1 3266 Intron NP_001275511.1
NM_133462.3 3266 Intron NP_597719.1
XM_011512439.2 3266 Intron XP_011510741.1
XM_017005739.1 3266 Intron XP_016861228.1
XM_017005740.1 3266 Intron XP_016861229.1

View Full Product Details