Product Details

SNP ID
rs74119748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:19816857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAGGGCTGCGAGTGTGGCAAGT[A/T]GCAAAGAGAGCCTCAGAGGTCCGAA
Phenotype
MIM: 606827
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PLXDC2 PubMed Links

Gene Details

Gene
PLXDC2
Gene Name
plexin domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282736.1 643 UTR 5 NP_001269665.1
NM_032812.8 643 UTR 5 NP_116201.7
XM_011519750.2 643 UTR 5 XP_011518052.1

View Full Product Details