Product Details

SNP ID
rs73421649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122841986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGGAGATACAGGTTAAATGGAG[C/T]GGAGCAGTTGCCCCTGACTTCCTGT
Phenotype
MIM: 605613
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HIP1R PubMed Links

Gene Details

Gene
HIP1R
Gene Name
huntingtin interacting protein 1 related
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303097.1 Intron NP_001290026.1
NM_001303099.1 Intron NP_001290028.1
NM_003959.2 Intron NP_003950.1
XM_011538963.1 Intron XP_011537265.1

View Full Product Details