Product Details

SNP ID
rs74026042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68269490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGTGTTATTGTTGCTATGGTTA[C/T]GTTAGGTGCACCAATGACTTCAAAT
Phenotype
MIM: 609362 MIM: 605641
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLA2G15 PubMed Links

Gene Details

Gene
PLA2G15
Gene Name
phospholipase A2 group XV
There are no transcripts associated with this gene.

Gene
SLC7A6
Gene Name
solute carrier family 7 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001076785.2 Intron NP_001070253.1
NM_003983.5 Intron NP_003974.3
XM_011523433.1 Intron XP_011521735.1
XM_011523434.1 Intron XP_011521736.1
XM_011523438.1 Intron XP_011521740.1
XM_011523439.1 Intron XP_011521741.1
XM_017023851.1 Intron XP_016879340.1

View Full Product Details