Product Details

SNP ID
rs73518694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17860405 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGCCGAAACGGGGAGCGTGAGG[C/T]TTAACGCTGTGGTACATACTGGATA
Phenotype
MIM: 604178
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPL18A PubMed Links

Gene Details

Gene
RPL18A
Gene Name
ribosomal protein L18a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000980.3 Intron NP_000971.1
Gene
SNORA68
Gene Name
small nucleolar RNA, H/ACA box 68
There are no transcripts associated with this gene.

View Full Product Details