Product Details

SNP ID
rs72951377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:104469307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCACTATGTATCAACGCCTCAA[G/A]TATTTACCATGATCTCCCCAAATGC
Phenotype
MIM: 611645
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CXXC4 PubMed Links

Gene Details

Gene
CXXC4
Gene Name
CXXC finger protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025212.3 4719 UTR 3 NP_079488.2
XM_011532284.2 4719 UTR 3 XP_011530586.1
XM_017008652.1 4719 UTR 3 XP_016864141.1

View Full Product Details