Product Details

SNP ID
rs72716481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:142023528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTTGAAAGTTTCATAGTTGTTT[A/C]ACAAAATATTTTTAATGTTGATAGT
Phenotype
MIM: 607494
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
INPP4B PubMed Links

Gene Details

Gene
INPP4B
Gene Name
inositol polyphosphate-4-phosphatase type II B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101669.1 8636 Intron NP_001095139.1
NM_003866.2 8636 Intron NP_003857.2
XM_005263323.3 8636 Intron XP_005263380.1
XM_005263325.3 8636 Intron XP_005263382.1
XM_011532391.2 8636 Intron XP_011530693.1
XM_011532392.2 8636 Intron XP_011530694.1
XM_017008794.1 8636 UTR 3 XP_016864283.1
XM_017008795.1 8636 Intron XP_016864284.1
XM_017008796.1 8636 Intron XP_016864285.1
XM_017008797.1 8636 Intron XP_016864286.1
XM_017008798.1 8636 Intron XP_016864287.1

View Full Product Details