Product Details

SNP ID
rs74112264
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:119032342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAATTAAAAAACAAATTATTGCT[G/A]GAACAGTTGGTGTTATCTCTTGGAA
Phenotype
MIM: 604733
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
WARS2 PubMed Links

Gene Details

Gene
WARS2
Gene Name
tryptophanyl tRNA synthetase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015836.3 1502 UTR 3 NP_056651.1
NM_201263.2 1502 UTR 3 NP_957715.1
XM_005270350.3 1502 UTR 3 XP_005270407.1
XM_011540494.2 1502 UTR 3 XP_011538796.1
XM_011540495.2 1502 UTR 3 XP_011538797.1
XM_017000038.1 1502 UTR 3 XP_016855527.1
XM_017000039.1 1502 UTR 3 XP_016855528.1
XM_017000040.1 1502 UTR 3 XP_016855529.1
XM_017000041.1 1502 UTR 3 XP_016855530.1
XM_017000042.1 1502 UTR 3 XP_016855531.1

View Full Product Details