Product Details

SNP ID
rs74084812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:51361569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGATATCCATCCCATAAAAATAAT[C/T]CTGAAAGTTACCTTCAATAAGGAAT
Phenotype
MIM: 604331
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INTS6 PubMed Links

Gene Details

Gene
INTS6
Gene Name
integrator complex subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039937.1 7461 UTR 3 NP_001035026.1
NM_001039938.1 7461 Intron NP_001035027.1
NM_001306091.1 7461 UTR 3 NP_001293020.1
NM_012141.2 7461 UTR 3 NP_036273.1
XM_011535040.2 7461 Intron XP_011533342.1
XM_011535041.1 7461 Intron XP_011533343.1
XM_017020540.1 7461 Intron XP_016876029.1
Gene
SERPINE3
Gene Name
serpin family E member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101320.1 7461 Intron NP_001094790.1
XM_017020707.1 7461 Intron XP_016876196.1

View Full Product Details