Product Details

SNP ID
rs74083708
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:68474887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGATAGTTAATAAGGCTCATCA[A/G]TTGTCCGTAATTTTGTTTTGGTTCA
Phenotype
MIM: 612002
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEPDC1 PubMed Links

Gene Details

Gene
DEPDC1
Gene Name
DEP domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114120.2 4703 UTR 3 NP_001107592.1
NM_017779.5 4703 UTR 3 NP_060249.2

View Full Product Details