Product Details

SNP ID
rs74101447
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:93152581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATATGGTAAGAAGTTACTGAATC[A/C]TAGGTTTAAGATCAGTGGCACAAAT
Phenotype
MIM: 616876
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMED5 PubMed Links

Gene Details

Gene
TMED5
Gene Name
transmembrane p24 trafficking protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167830.1 3275 UTR 3 NP_001161302.1
NM_016040.4 3275 UTR 3 NP_057124.3

View Full Product Details