Product Details

SNP ID
rs73013906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125498142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACACATATATATGTCATATATA[T/C]GTCTCATATAGATGTCCCATATATA
Phenotype
MIM: 604825
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
FEZ1 PubMed Links

Gene Details

Gene
FEZ1
Gene Name
fasciculation and elongation protein zeta 1
There are no transcripts associated with this gene.

Gene
LOC403312
Gene Name
putative uncharacterized protein MGC39545
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301851.1 2037 UTR 3 NP_001288780.1

View Full Product Details