Product Details

SNP ID
rs73258310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:114061831 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGACATTAGGTTGATTGTCTATGC[A/T]TGTGCAACTGGGAAGTGGAGGATCT
Phenotype
MIM: 605879
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNN2 PubMed Links

Gene Details

Gene
KCNN2
Gene Name
potassium calcium-activated channel subfamily N member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278204.1 Intron NP_001265133.1
NM_021614.3 Intron NP_067627.2
NM_170775.2 Intron NP_740721.1
XM_011543387.1 Intron XP_011541689.1
XM_011543389.1 Intron XP_011541691.1
XM_017009457.1 Intron XP_016864946.1

View Full Product Details