Product Details

SNP ID
rs73088167
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:206066173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCTGGTCTTTAGAGGCGTATGC[C/T]TCTGGTTCTACCACTCTCTGCAGTC
Phenotype
MIM: 616088
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf186 PubMed Links

Gene Details

Gene
C1orf186
Gene Name
chromosome 1 open reading frame 186
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007544.3 Intron NP_001007545.1
XM_005272738.3 Intron XP_005272795.1
Gene
LOC100505650
Gene Name
uncharacterized LOC100505650
There are no transcripts associated with this gene.

View Full Product Details