Product Details

SNP ID
rs76307406
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:160488371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAAGGGAATCCAAGATTTCTAG[A/C]TAGATCATTGCTTACATCATTTTGA
Phenotype
MIM: 606446
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLAMF6 PubMed Links

Gene Details

Gene
SLAMF6
Gene Name
SLAM family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184714.1 Intron NP_001171643.1
NM_001184715.1 Intron NP_001171644.1
NM_001184716.1 Intron NP_001171645.1
NM_052931.4 Intron NP_443163.1
XM_017000215.1 Intron XP_016855704.1
XM_017000216.1 Intron XP_016855705.1
XM_017000217.1 Intron XP_016855706.1

View Full Product Details