Product Details

SNP ID
rs78500642
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:161099656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTTGCAAGGCTTGTGAGCAGTG[G/T]GCAGGGGTCCCAGAAGGGGCCCCAT
Phenotype
MIM: 613466
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KLHDC9 PubMed Links

Gene Details

Gene
KLHDC9
Gene Name
kelch domain containing 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007255.2 891 Missense Mutation TGG,TGT W255C NP_001007256.1
NM_152366.4 891 Missense Mutation GGG,GTG G249V NP_689579.3
Gene
PFDN2
Gene Name
prefoldin subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012394.3 891 Intron NP_036526.2
XM_011509624.2 891 Intron XP_011507926.1

View Full Product Details