Product Details

SNP ID
rs76677462
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:166612496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCAAAAAGCGATTGCTGTGATT[G/T]GAGCTGGAATTAGCGGACTGGGGGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FMO9P PubMed Links

Gene Details

Gene
FMO9P
Gene Name
flavin containing monooxygenase 9 pseudogene
There are no transcripts associated with this gene.

View Full Product Details