Product Details

SNP ID
rs74899700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:169067818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTAACATGACTTCATTAACTGATC[A/G]CCTATGTGGGACCATTGGACAAGGA
Phenotype
MIM: 612131
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHRS9 PubMed Links

Gene Details

Gene
DHRS9
Gene Name
dehydrogenase/reductase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142270.1 Intron NP_001135742.1
NM_001142271.1 Intron NP_001135743.1
NM_001289763.1 Intron NP_001276692.1
NM_199204.1 Intron NP_954674.1
XM_011510475.1 Intron XP_011508777.1

View Full Product Details