Product Details

SNP ID
rs79465025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:178201858 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGTGTGCCTGACAGTGTTCCC[A/G]TGACCATTCTTACATGTGCTTGTTA
Phenotype
MIM: 606734
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OSBPL6 PubMed Links

Gene Details

Gene
OSBPL6
Gene Name
oxysterol binding protein like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201480.1 Intron NP_001188409.1
NM_001201481.1 Intron NP_001188410.1
NM_001201482.1 Intron NP_001188411.1
NM_032523.3 Intron NP_115912.1
NM_145739.2 Intron NP_665682.1
XM_011510539.1 Intron XP_011508841.1
XM_017003265.1 Intron XP_016858754.1
XM_017003266.1 Intron XP_016858755.1
XM_017003267.1 Intron XP_016858756.1
XM_017003268.1 Intron XP_016858757.1
XM_017003269.1 Intron XP_016858758.1
XM_017003270.1 Intron XP_016858759.1
XM_017003271.1 Intron XP_016858760.1
XM_017003272.1 Intron XP_016858761.1
XM_017003273.1 Intron XP_016858762.1
XM_017003274.1 Intron XP_016858763.1
XM_017003275.1 Intron XP_016858764.1
XM_017003276.1 Intron XP_016858765.1
XM_017003277.1 Intron XP_016858766.1

View Full Product Details