Product Details

SNP ID
rs75065977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218452166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGATCAAAAGGTTGTATCTAGAA[C/T]TAATTGCCATCAAGTTCCAATTCAA
Phenotype
MIM: 193040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP37 PubMed Links

Gene Details

Gene
USP37
Gene Name
ubiquitin specific peptidase 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020935.2 6081 UTR 3 NP_065986.2
XM_005246720.2 6081 UTR 3 XP_005246777.1
XM_005246721.3 6081 UTR 3 XP_005246778.1
XM_005246722.3 6081 UTR 3 XP_005246779.1
XM_005246724.1 6081 UTR 3 XP_005246781.1
XM_011511538.2 6081 UTR 3 XP_011509840.1
XM_011511541.2 6081 Intron XP_011509843.1
XM_017004577.1 6081 UTR 3 XP_016860066.1
Gene
VIL1
Gene Name
villin 1
There are no transcripts associated with this gene.

View Full Product Details