Product Details

SNP ID
hCV1073606
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:108307971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGCTTGCTGTTGTGGACTACATG[A/T]GAAGACAAAAGAGGTAATGTAATGA
Phenotype
MIM: 607585
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ATM PubMed Links

Gene Details

Gene
ATM
Gene Name
ATM serine/threonine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000051.3 6134 Nonsense Mutation AGA,TGA R1917* NP_000042.3
Gene
C11orf65
Gene Name
chromosome 11 open reading frame 65
There are no transcripts associated with this gene.

View Full Product Details