Product Details
- SNP ID
-
hCV1094460
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:154379709 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCAGGATCAACTCGTAGGCTTTA[C/G]GAGAAGAAGATCTTCGAGTACGAGA
- Phenotype
-
MIM: 300384
MIM: 300017
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
EMD
PubMed Links
Gene Details
- Gene
- EMD
- Gene Name
- emerin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000117.2 |
350 |
Nonsense Mutation |
TAC,TAG |
Y34* |
NP_000108.1 |
- Gene
- FLNA
- Gene Name
- filamin A
There are no transcripts associated with this gene.
- Gene
- LOC107987332
- Gene Name
- uncharacterized LOC107987332
There are no transcripts associated with this gene.
View Full Product Details