Product Details

SNP ID
rs28405099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41139883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCAAGGAGCTCAGCAGCAAGAG[A/G]AGGCACAGCACAAGGGACGGGGGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRTAP4-5 PubMed Links

Gene Details

Gene
KRTAP4-5
Gene Name
keratin associated protein 4-5
There are no transcripts associated with this gene.

Gene
KRTAP4-6
Gene Name
keratin associated protein 4-6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030976.1 605 Missense Mutation TCC,TTC S202F NP_112238.1

View Full Product Details