Product Details

SNP ID
rs655681
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:127698625 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACAAGCAACAGCATATTCACTCAG[A/G]TGAAACAATCATACAGGACCTATTC
Phenotype
MIM: 613607
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
THEMIS PubMed Links

Gene Details

Gene
THEMIS
Gene Name
thymocyte selection associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010923.2 Intron NP_001010923.1
NM_001164685.1 Intron NP_001158157.1
NM_001164687.1 Intron NP_001158159.1
NM_001318531.1 Intron NP_001305460.1
XM_011535813.2 Intron XP_011534115.1
XM_011535814.2 Intron XP_011534116.1
XM_011535816.1 Intron XP_011534118.1
XM_017010848.1 Intron XP_016866337.1
XM_017010849.1 Intron XP_016866338.1

View Full Product Details