Product Details

SNP ID
rs35590990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:134070302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTAACCAGACCAGGACCTTTTTC[C/G]GTTCTTAGATTATTTTATTGTTTGA
Phenotype
MIM: 606871
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
JAM3 PubMed Links

Gene Details

Gene
JAM3
Gene Name
junctional adhesion molecule 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205329.1 Intron NP_001192258.1
NM_032801.4 Intron NP_116190.3

View Full Product Details