Product Details
- SNP ID
-
rs1043327
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:18444162 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCTTGCTGGAGCAGAGACCCTGCCC[A/G]AGGAGGAGAGGCTGGGGAAAGGGGT
- Phenotype
-
MIM: 600284
MIM: 611670
MIM: 607391
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ELL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80253247] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ELL
- Gene Name
- elongation factor for RNA polymerase II
- Gene
- ISYNA1
- Gene Name
- inositol-3-phosphate synthase 1
There are no transcripts associated with this gene.
- Gene
- SSBP4
- Gene Name
- single stranded DNA binding protein 4
There are no transcripts associated with this gene.
View Full Product Details