Product Details

SNP ID
rs1566289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:73242634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGCCTCCTCCCAAAGCCAGAAA[A/G]ACTCTAAACCTCAAGTTAACCAGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C17orf80 PubMed Links

Gene Details

Gene
C17orf80
Gene Name
chromosome 17 open reading frame 80
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100621.2 Intron NP_001094091.1
NM_001100622.2 Intron NP_001094092.1
NM_001288770.1 Intron NP_001275699.1
NM_001288771.1 Intron NP_001275700.1
NM_017941.5 Intron NP_060411.2
XM_005257487.3 Intron XP_005257544.1
XM_006721966.3 Intron XP_006722029.1
XM_011524961.1 Intron XP_011523263.1
XM_011524962.2 Intron XP_011523264.1
XM_017024806.1 Intron XP_016880295.1
Gene
CPSF4L
Gene Name
cleavage and polyadenylation specific factor 4 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129885.1 Intron NP_001123357.1
XM_011525115.2 Intron XP_011523417.1

View Full Product Details