Product Details

SNP ID
rs282584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:112695505 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTGCTTTTTGGAACCCTGTTGAAT[A/G]TCTGCTTATTAGGAACAAGCATCTT
Phenotype
MIM: 605868
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP11A PubMed Links

Gene Details

Gene
ATP11A
Gene Name
ATPase phospholipid transporting 11A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015205.2 Intron NP_056020.2
NM_032189.3 Intron NP_115565.3
XM_005268299.4 Intron XP_005268356.1
XM_005268300.4 Intron XP_005268357.1
XM_005268301.4 Intron XP_005268358.1
XM_005268303.4 Intron XP_005268360.1
XM_005268305.4 Intron XP_005268362.1
XM_005268306.4 Intron XP_005268363.1
XM_011537480.2 Intron XP_011535782.1
XM_017020490.1 Intron XP_016875979.1
XM_017020491.1 Intron XP_016875980.1
XM_017020492.1 Intron XP_016875981.1

View Full Product Details