Product Details

SNP ID
rs12001254
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34813038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATATAAGAAGCCAGTGAAAGCAC[A/G]TAAAGATACTTTACAGTTAACATGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHF24 PubMed Links

Gene Details

Gene
PHF24
Gene Name
PHD finger protein 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304333.2 Intron NP_001291262.1
NM_015297.2 Intron NP_056112.1
XM_011517827.1 Intron XP_011516129.1
XM_011517828.2 Intron XP_011516130.1
XM_011517829.1 Intron XP_011516131.1
XM_017014553.1 Intron XP_016870042.1
XM_017014554.1 Intron XP_016870043.1
XM_017014555.1 Intron XP_016870044.1
XM_017014556.1 Intron XP_016870045.1

View Full Product Details