Product Details

SNP ID
rs35256639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:68934209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTACAAAAGCTCTTATTTCTAG[A/G]TTGGTATTATAAGAAGGCTCAATGT
Phenotype
MIM: 606572 MIM: 609399
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NOX5 PubMed Links

Gene Details

Gene
NOX5
Gene Name
NADPH oxidase, EF-hand calcium binding domain 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184779.1 Intron NP_001171708.1
NM_001184780.1 Intron NP_001171709.1
NM_024505.3 Intron NP_078781.3
Gene
SPESP1
Gene Name
sperm equatorial segment protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145658.3 Intron NP_663633.1

View Full Product Details