Product Details

SNP ID
rs8031179
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65659696 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGAACAGGTTTACCACTTTGTATT[C/T]GTGTTTTAGGTTTTAAGACTAAATC
Phenotype
MIM: 600382 MIM: 603617
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DENND4A PubMed Links

Gene Details

Gene
DENND4A
Gene Name
DENN domain containing 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144823.2 8125 UTR 3 NP_001138295.1
NM_001320835.1 8125 UTR 3 NP_001307764.1
NM_005848.4 8125 UTR 3 NP_005839.3
XM_005254120.4 8125 UTR 3 XP_005254177.1
XM_005254121.3 8125 UTR 3 XP_005254178.1
XM_005254122.4 8125 UTR 3 XP_005254179.1
XM_006720371.3 8125 UTR 3 XP_006720434.1
XM_011521156.2 8125 Intron XP_011519458.1
XM_011521157.2 8125 UTR 3 XP_011519459.1
XM_017021862.1 8125 UTR 3 XP_016877351.1
XM_017021863.1 8125 UTR 3 XP_016877352.1
XM_017021864.1 8125 Intron XP_016877353.1
XM_017021865.1 8125 UTR 3 XP_016877354.1
Gene
SLC24A1
Gene Name
solute carrier family 24 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001254740.1 8125 Intron NP_001241669.1
NM_001301031.1 8125 Intron NP_001287960.1
NM_001301032.1 8125 Intron NP_001287961.1
NM_001301033.1 8125 Intron NP_001287962.1
NM_004727.2 8125 Intron NP_004718.1
XM_005254778.3 8125 Intron XP_005254835.1
XM_005254781.3 8125 Intron XP_005254838.1
XM_006720768.3 8125 Intron XP_006720831.1
XM_011522219.1 8125 Intron XP_011520521.1
XM_011522220.2 8125 Intron XP_011520522.1
XM_011522221.2 8125 Intron XP_011520523.1
XM_011522222.2 8125 Intron XP_011520524.1
XM_011522226.2 8125 Intron XP_011520528.1
XM_017022724.1 8125 Intron XP_016878213.1
XM_017022725.1 8125 Intron XP_016878214.1

View Full Product Details