Product Details

SNP ID
rs7198357
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:67850716 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGCATGCGGTGGTTGGGATATTGC[C/T]GCACAAGGATGAGGCAGGATACAAG
Phenotype
MIM: 611510 MIM: 605813 MIM: 609119
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CENPT PubMed Links

Gene Details

Gene
CENPT
Gene Name
centromere protein T
There are no transcripts associated with this gene.

Gene
NUTF2
Gene Name
nuclear transport factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322038.1 Intron NP_001308967.1
NM_001322039.1 Intron NP_001308968.1
NM_001322040.1 Intron NP_001308969.1
NM_001322041.1 Intron NP_001308970.1
NM_005796.2 Intron NP_005787.1
Gene
THAP11
Gene Name
THAP domain containing 11
There are no transcripts associated with this gene.

View Full Product Details