Product Details
- SNP ID
-
rs11848712
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:89955818 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCCCCGTCCTGGGTAAACACCCG[A/G]CGGGCTTTAGGACCCAGCGGTGGGC
- Phenotype
-
MIM: 607198
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EFCAB11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113492506] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EFCAB11
- Gene Name
- EF-hand calcium binding domain 11
There are no transcripts associated with this gene.
- Gene
- TDP1
- Gene Name
- tyrosyl-DNA phosphodiesterase 1
View Full Product Details